Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ro 04-5595 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
64047-73-0
C19H23Cl2NO2
368.298
NVIPBLQAFKRFSZ-UHFFFAOYSA-N
ro 04-5595 hydrochloride, 1-2-4-chlorophenyl ethyl-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-3,4-dihydro-1h-isoquinolin-7-ol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-7-ol hydrochloride
16759151
1-[2-(4-chlorophenyl)ethyl]-6-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-7-ol;hydrochloride
CN1CCC2=CC(=C(C=C2C1CCC3=CC=C(C=C3)Cl)O)OC.Cl
This item is not returnable. View return policy
64047-73-0
C19H23Cl2NO2
368.298
NVIPBLQAFKRFSZ-UHFFFAOYSA-N
ro 04-5595 hydrochloride, 1-2-4-chlorophenyl ethyl-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-3,4-dihydro-1h-isoquinolin-7-ol hydrochloride, 1-2-4-chlorophenyl ethyl-6-methoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-7-ol hydrochloride
16759151
1-[2-(4-chlorophenyl)ethyl]-6-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-7-ol;hydrochloride
CN1CCC2=CC(=C(C=C2C1CCC3=CC=C(C=C3)Cl)O)OC.Cl

GluN2B-selective NMDA antagonist

Selective antagonist for GluN2B (formally NR2B) containing NMDA receptors (Ki = 31 nM).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Quantity 50 mg
Formula Weight 368.3
Percent Purity >98%
Chemical Name or Material Ro 04-5595 hydrochloride

Recommended Storage : Desiccate at +4°C