Ro 04-5595 hydrochloride, Tocris

Selective NR2B antagonist

Manufacturer: R&D Systems 2005/10

Quantity 10mg

 View more versions of this product

Catalog No. 20-051-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C19H22ClNO2·HCl
    Formula Weight 368.3
    Synonym 1-[2-(4-Chlorophenyl)ethyl]-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride
    Chemical Name or Material Ro 04-5595 hydrochloride

    Specifications

    Formula Weight 368.3
    Quantity 10mg
    Synonym 1-[2-(4-Chlorophenyl)ethyl]-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride
    Molecular Formula C19H22ClNO2·HCl
    CAS 64047-73-0
    Recommended Storage Desiccate at +4°C
    Assay Percent Range >98%
    Chemical Name or Material Ro 04-5595 hydrochloride

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit