Learn More
Ro 10-5824 dihydrochloride, Tocris
Selective D4 receptor partial agonist
Manufacturer:  R&D Systems 2329/50
Quantity 50mg
 View more versions of this product
Catalog No. 23-295-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
353.29
Ro 10-5824 dihydrochloride
C17H20N4·2HCl
5-[(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)methyl]-2-methyl-4-pyrimidinamine dihydrochloride
Specifications
50mg
>99%
353.29
Ro 10-5824 dihydrochloride
189744-94-3
C17H20N4·2HCl
5-[(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)methyl]-2-methyl-4-pyrimidinamine dihydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature