Ro 106-9920, Tocris

Inhibitor of NF-κB activation

Manufacturer: R&D Systems 1778/10

Quantity 10mg

 View more versions of this product

Catalog No. 17-781-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C10H7N5OS
    Formula Weight 245.26
    Synonym 6-(Phenylsulfinyl)tetrazolo[1, 5-b]pyridazine
    Chemical Name or Material Ro 106-9920

    Specifications

    Formula Weight 245.26
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C10H7N5OS
    CAS 62645-28-7
    Synonym 6-(Phenylsulfinyl)tetrazolo[1, 5-b]pyridazine
    Recommended Storage Store at +4°C
    Chemical Name or Material Ro 106-9920

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit