Learn More
Tocris Bioscience™ Ro 25-6981 maleate
GluN2B-selective NMDA antagonist
$124.12 - $1085.76
Specifications
Chemical Name or Material | (αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate |
---|---|
CAS | 1312991-76-6 |
Assay Percent Range | >98% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C22H29NO2.C4H4O4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
15-941-0
|
Tocris Bioscience™
1594/10 |
10 mg |
Each for $263.32
|
|
|||||
15-945-0
|
Tocris Bioscience™
1594/50 |
50 mg |
Each for $1,085.76
|
|
|||||
15-941-R
|
Tocris Bioscience™
1594/1 |
1 mg |
Each for $124.12
|
|
|||||
Description
Potent and selective activity-dependent blocker of NMDA receptors containing the GluN2B (formally NR2B) subunit. IC50 values are 0.009 and 52 μM for cloned receptor subunit combinations GluN1C/GluN2B and GluN1C/GluN2A respectively. Displays neuroprotectant effects in vivo and in vitro.Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
(αR,βS)-α-(4-Hydroxyphenyl)-β-methyl-4-(phenylmethyl)-1-piperidinepropanol maleate | |
>98% | |
C22H29NO2.C4H4O4 |
1312991-76-6 | |
Soluble to 100mM in DMSO | |
Observed MW: 455.55 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.