Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ro 67-4853, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Positive allosteric modulator of mGlu group I receptors (pEC50 value is 7.16 for the rat mGlu1a receptor). Exhibits activity at all group I receptors, including human and rat mGlu1 and rat mGlu5. Enhances the effects of (S)-DHPG in CA3 neurons (EC50 = 95 nM).
Specifications
Quantity | 10 mg |
Formula Weight | 325.36 |
Percent Purity | >99% |
Chemical Name or Material | Ro 67-4853 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction