Learn More
Ro 8-4304 hydrochloride, Tocris
Selective NR2B antagonist
Manufacturer:  R&D Systems 3343/50
Quantity 50mg
 View more versions of this product
Catalog No. 33-435-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
406.88
Ro 8-4304 hydrochloride
C21H23FN2O3·HCl
4-[3-[4-(4-Fluorophenyl)-1,2,3,6-tetrahydro-1(2H)-pyridinyl]-2-hydroxypropoxy]benzamide hydrochloride
Specifications
50mg
>99%
406.88
Ro 8-4304 hydrochloride
1312991-77-7
C21H23FN2O3·HCl
4-[3-[4-(4-Fluorophenyl)-1,2,3,6-tetrahydro-1(2H)-pyridinyl]-2-hydroxypropoxy]benzamide hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature