Learn More
ROBO4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 202211AP
Catalog No. 50-173-2776
Add to cart
Description
Description
This antibody is specific to ROBO4.
Roundabouts (ROBO) are cell-surface receptors that mediate repulsive signaling mechanisms at the central nervous system midline. However, ROBOs may also mediate attraction mechanisms in the context of vascular development. ROBO4 is a novel roundabout protein which is restricted in expression to endothelial cells in vitro and sites of angiogenesis in vivo. Because it is not expressed on normal endothelial cells, in vivo, it is a promising tumor endothelial marker.
Specifications
Specifications
ROBO4
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C310, Q8WZ75
ECSM4, Magic roundabout, MRB, ROBO4, Roundabout homolog 4
Rabbit
IgG
150 μL
-20°C
Primary
54538, 74144
Western Blot
Unconjugated
ROBO4
Liquid
ROBO4
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse