Learn More
ROBO4, Rat anti-Mouse, Alexa Fluor 405, Clone: 274914, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB50041V100UG
Catalog No. FAB50041V10
Add to cart
This item is not returnable. View return policy
Description
Description
ROBO4 Monoclonal specifically detects ROBO4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
ROBO4
274914
Flow Cytometry 0.25-1 ug/10^6 cells
Q8C310
ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila)
Y3 rat myeloid cell line transfected with mouse ROBO4, Leu28-Ser1012, Accession # Q8C310
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
54538
Mouse
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Rat
IgG2a
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects mouse ROBO4 in direct ELISAs.