Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AZER Scientific Robotic Conductive Tips (Hamilton Compatible)
SDP

Catalog No. p-7112444
Click to view available options
Volume (Metric):
10 μL
300 μL
Tip Style:
Automation
Filtered:
Filtered
Non-filtered
Format:
Blister Tray
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to be compatible with Hamilton Microlab Star™, Nimbus™ Vantage™ pipetting instruments.

Conductivity enables the system to recognize the filling height to assure minimal immersion of the tip into the liquid.

  • Manufactured under cleanroom conditions
  • Each lot tested according to strict quality management system to ensure reproducible results for every application
  • High quality, tested by independent labs to be free of DNase, RNase, ATP and pyrogens
  • Conductive tips allow pipettor to sense liquid levels
  • Made from pure Polypropylene

Specifications

Volume (Metric) 300 μL
Filtered Filtered
Sterility Non-sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase, ATP, and Pyrogen Free
For Use With (Application) Pipetting and dispensing
Format Blister Tray
Tip Style Automation
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB Nimbus, Hamilton MICROLAB Vantage
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.