Learn More
SP Bel-Art Robotic Sterile Pipette Tips
Designed for use with automated liquid handling equipment
$343.50 - $631.50
Specifications
Certifications/Compliance CE marked
Filtered Filtered
Sterility Sterile
Disposable Yes
For Use With (Application) For automated liquid handling equipment
View More Specs
Products 8
Catalog Number Mfr. No. Conductivity Volume (Metric) For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. Conductivity Volume (Metric) For Use With (Equipment) Material Price Quantity & Availability  
03-411-871
View Documents
SP Bel-Art
F378603005
Yes 50 μL Hamilton™ STAR™, STARlet and NIMBUS™ Carbon filled polypropylene
Case of 2304 for $631.50
Add to cart
03-411-872
View Documents
SP Bel-Art
F378603100
Yes 1000 μL Hamilton™ STAR™, STARlet and NIMBUS™ Carbon filled polypropylene
Case of 1536 for $458.00
Add to cart
03-411-867
View Documents
SP Bel-Art
F378601005
No 50 μL Hamilton™ STAR™, STARlet and NIMBUS™ Clear polypropylene
Case of 2304 for $531.00
Add to cart
03-411-868
View Documents
SP Bel-Art
F378601100
No 1000 μL Hamilton™ STAR™, STARlet and NIMBUS™ Clear polypropylene
Case of 1536 for $393.50
Add to cart
03-411-873
View Documents
SP Bel-Art
F378604005
Yes 50 μL Tecan™ Genesis Freedom™, Freedom EVO™ and Miniprep with LiHa Carbon filled polypropylene
Case of 2304 for $604.00
Add to cart
03-411-874
View Documents
SP Bel-Art
F378604100
Yes 1000 μL Tecan™ Genesis Freedom™, Freedom EVO™ and Miniprep with LiHa Carbon filled polypropylene
Case of 1536 for $453.00
Add to cart
03-411-869
View Documents
SP Bel-Art
F378602005
No 50 μL Tecan™ Genesis Freedom™, Freedom EVO™ and Miniprep with LiHa Clear polypropylene
Case of 2304 for $471.50
Add to cart
03-411-870
View Documents
SP Bel-Art
F378602100
No 1000 μL Tecan™ Genesis Freedom™, Freedom EVO™ and Miniprep with LiHa Clear polypropylene
Case of 1536 for $343.50
Add to cart
Description
Description
  • Sterile, disposable, filtered pipette tips can be used in robotic systems
    • Filtered to prevent contamination of pipette arm
  • Conductive tip models detect liquid level when pipetting conductive liquids
    • Ensures repeatable volume accuracy
  • DNase, RNase, pyrogen free, manufactured in Class 8 clean room
  • CE marked
Specifications
Specifications
CE marked
Sterile
For automated liquid handling equipment
Filtered
Yes
Robotic pipette tips