Learn More
Fisherbrand™ Lab Automation Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory procedures
Catalog No. 02-707-340
Add to Cart
Description
Description
Provide maximum performance, accuracy and reproducibility in laboratory procedures.
  • Non-Sterile
  • Polypropylene
  • Natural color
  • Rack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 μL
Natural
Ideal for demanding HTS procedures, provide maximum performance, accuracy and reproducibility in lab automation procedures
Automation
BioTek Precision, Tecan TeMo, Tecan GenMate
Non-sterile
Autoclavable
Rack
Single-use
Polypropylene
Product Suggestions
Product Suggestions