Learn More
Selleck Chemical LLCSupplier Diversity Partner ROC-325 DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S85275MG
ROC-325 is an orally available novel inhibitor of lysosomal-mediated autophagy which diminishes AML cell viability with the IC50 range of 0.7-2.2 μM.
Catalog No. 50-194-8339