Learn More
Roche Diagnostics CoaguChek™ S Prothrombin Time Monitoring System
PT results in one minute; CLIA Waived
$51.20
Specifications
For Use With (Equipment) CoaguChek XS System
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
23-312188
View Documents
Roche Diagnostics
11621173001
Capillary Tubes and Bulbs
Pack of 100 for $51.20
Add to cart
22-045-625
View Documents
Roche Diagnostics
04348150001
Lancets This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
These Accessories are for use with the CoaguChek XS System.
The CoaguChek XS System (04837975001) is a portable, extremely efficient instrument used by doctors and healthcare professionals to monitor patients under anticoagulant therapy. The System is less invasive, improves patient compliance and offers the advantage of immediate therapy adjustment.The CoaguChek S System tests for PT levels. Simply turn on the monitor, insert test strip and apply a sample from a fingerstick. Results appear within one minute on the LCD screen and can be displayed as INR, INR and Seconds, INR and % Quick or INR and Ratio.
  • Memory stores up to 60 test results
  • Monitor is easy-to-use; users are guided through all testing steps and settings menus by universal icon, ensuring correct operation
  • No calibration, pipetting or reagent preparation is required
  • CLIA-waived, perfect for in-office or on-site testing
  • Provides consistent results test-to-test, lot-to-lot, monitor-to-monitor
  • System check is performed every time the monitor is turned on and performance can be verified with either electronic or liquid controls
Order Info
Roche provides technical assistance 24 hours-a-day.
Specifications
Specifications
CoaguChek XS System