Learn More
Roche Diagnostics Glucose Test Strips for Accutrend Plus™ Meters
For the determination of glucose in capillary whole blood
Manufacturer:  Roche Diagnostics 11447475160
Catalog No. 22-045-871
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Can be dosed outside of the meter to prevent cross-contamination for community health events
Test strips contain:
  • Yellow protective mesh, impregnated with a surfactant (application field)
  • Glass fiber fleece that works as a separating layer for blood cells
  • Reaction film in which the color formation takes place
  • Bar code on the reverse side, which is read by the meter to confirm test strip identity (lot number on the test strip is congruent with the stored code)
Specifications
Specifications
Accutrend Plus™ system
18 Months
20 to 600mg/dL
25 Vials
12 sec.
Room Temperature
STD = 1.8mg/dL
A hanging of drop of blood