Roche MagNA Pure LC Disposable Plastic Accessories
The MagNA Pure LC disposable plastic accessories meet stringent requirements for quality and convenience for the safety of samples
Manufacturer: Roche Diagnostics 03004066001
 View more versions of this product
Catalog No. 50-720-3005
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For storing up to 3.5mL of MagNA pure LC reagents on the stage of the MagNA pure LC instrument
    For Use With (Equipment) Roche MagNA Pure LC Total Nucleic Acid Isolation Kit
    Product Type Reagent Tub, Small
    Product Name Reagent Tub, Small
  • Description & Specifications
Related Products
Product Content Feedback