Learn More
Roche Diagnostics MagNA Pure LC Disposable Plastic Accessories
The MagNA Pure LC disposable plastic accessories meet stringent requirements for quality and convenience for the safety of samples
Manufacturer: Roche Diagnostics 03004074001
Description
- Inert to most chemicals used in life-science laboratories
- Free of nucleases and amplification inhibitors (for PCR/RT-PCR
Specifications
For use with MagNA pure LC large reagent tubs | |
Roche MagNA Pure LC Total Nucleic Acid Isolation Kit | |
Tub Lids, Large |