Learn More
Roche Diagnostics MagNA Pure LC Disposable Plastic Accessories Non-distribution product as customer accommodation.
The MagNA Pure LC disposable plastic accessories meet stringent requirements for quality and convenience for the safety of samples
Manufacturer:  Roche Diagnostics 03004082001
 View more versions of this product
Catalog No. 50-720-3007
Add to cart
Description
Description
  • Inert to most chemicals used in life-science laboratories
  • Free of nucleases and amplification inhibitors (for PCR/RT-PCR
Specifications
Specifications
Roche MagNA Pure LC Total Nucleic Acid Isolation Kit
For use with MagNA pure LC small and medium reagent tubs
Tub Lids, Small/Medium