Learn More
Roche Diagnostics MagNA Pure LC Disposable Plastic Accessories Non-distribution product as customer accommodation.
The MagNA Pure LC disposable plastic accessories meet stringent requirements for quality and convenience for the safety of samples
$167.68 - $415.40
Specifications
For Use With (Equipment) Roche MagNA Pure LC Total Nucleic Acid Isolation Kit
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
50-720-3051 Non-distribution product as customer accommodation.
View Documents
Roche Diagnostics
03118827001
For sealing the MagNA pure LC sample cartridge for storage purposes
Each for $167.68
Add to cart
50-720-3007 Non-distribution product as customer accommodation.
View Documents
Roche Diagnostics
03004082001
For use with MagNA pure LC small and medium reagent tubs
Each for $415.40
Add to cart
50-720-3006 Non-distribution product as customer accommodation.
View Documents
Roche Diagnostics
03004074001
For use with MagNA pure LC large reagent tubs
Each for $203.36
Add to cart
50-720-3005 Non-distribution product as customer accommodation.
View Documents
Roche Diagnostics
03004066001
For storing up to 3.5mL of MagNA pure LC reagents on the stage of the MagNA pure LC instrument
Each for $234.36
Add to cart
Description
Description
  • Inert to most chemicals used in life-science laboratories
  • Free of nucleases and amplification inhibitors (for PCR/RT-PCR
Specifications
Specifications
Roche MagNA Pure LC Total Nucleic Acid Isolation Kit