Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Rochester-Pean Forceps, Sklar, 7-1/4 in., Blunt, Curved, Serrated, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21485
Econo Rochester-Pean Forceps, Sklar, 7-1/4 in., Blunt, Curved, Serrated, Stainless Steel. Econo Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with full, horizontally serrated jaws, and a length of 7-1/4 inches. It is sold in boxes of 12.
Catalog No. 50-143-7646