Learn More
Sklar InstrumentsSupplier Diversity Partner Rochester-Pean Forceps, Sklar, 8 in., Straight, Serrated, Stainless Steel. Sklar Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 172480
Rochester-Pean Forceps, Sklar, 8 in., Straight, Serrated, Stainless Steel. Sklar Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. This product is straight with full, horizontally serrated jaws, and a length of 8 inches.
Catalog No. 50-143-7401