Learn More
Sklar InstrumentsSupplier Diversity Partner Rochester-Ochsner (Kocher) Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Curved, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 172655
Rochester-Ochsner (Kocher) Forceps, Sklar, 5-1/2 in., 1x2 Teeth, Curved, Serrated, Stainless Steel. Sklar Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels. They have horizontal serrations the entire length of the jaw as well as 1 X 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. This product is curved with full, horizontally serrated jaws, and a length of 5-1/2 inches.
Catalog No. 50-143-7402