Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Sklar InstrumentsSupplier Diversity Partner Forceps, Surgi-OR Rochester-Pean Hemostatic Forceps, Sklar 5-1/2 in. Straight, Serrated, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 95470
Forceps, Surgi-OR Rochester-Pean Hemostatic Forceps, Sklar, 5-1/2 in. Straight, Serrated, Stainless Steel, EA. Surgi-OR Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with full, horizontally serrated jaws, and a length of 5-1/2 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6518