Learn More
Sklar InstrumentsSupplier Diversity Partner Sklarlite Rochester-Pean Forceps, Sklar, 6-1/4 in., Blunt, Straight, Serrated, Extra Light (SklarLite), Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 232532
Sklarlite Rochester-Pean Forceps, Sklar, 6-1/4 in., Blunt, Straight, Serrated, Extra Light (SklarLite), Stainless Steel. Sklarlite Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Sklarlite instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue. This product is straight with full, horizontally serrated jaws, and a length of 6-1/4 inches.
Catalog No. 50-143-7983