Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Rochester-Pean Forceps, Sklar, 7-1/4 in., Curved, Serrated, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 902292
Sklar Black Rochester-Pean Forceps, Sklar, 7-1/4 in., Curved, Serrated, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis during laser procedures. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.
Catalog No. 50-143-6568