Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Surgi-OR Rochester-Pean Hemostatic Forceps, Sklar 9 in., Straight, Serrated, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 95483
Forceps, Surgi-OR Rochester-Pean Hemostatic Forceps, Sklar, 9 in., Straight, Serrated, Stainless Steel, EA.Surgi-OR Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis. The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with full, horizontally serrated jaws, and a length of 9 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6525