Learn More
American Educational Products Rock Cycle Classroom Project
Millions of years of geological processes captured in one chart.
Manufacturer: American Educational Products 2521
Includes: Complete kit includes wall chart, 30 student charts, 30 specimens of each rock on the chart, test kit and Scott Resources Rocks and Minerals Study Guide
Description
- Covers geological concepts such as weathering of rocks near the Earth's surface; metamorphic transformation by pressure, heat and water; and magmatic assimilation
- Examine 12 rock specimens and identify each sample as an igneous, sedimentary or metamorphic rock
- Wood-framed wall chart comes complete with mounted specimens and measures 18 x 24 in. (45.7 x 61.0cm); also available as a handy notebook reference sheet
Specifications
6 to 12 | |
Provides an introductory study of the interaction of igneous, sedimentary, and metamorphic rocks | |
Rock cycle chart |