Learn More
Rock and Mineral Test Kit
Test to distinguish between rocks and minerals.
Specifications
For Use With (Application) | Used to conduct a series of tests to distinguish between rocks and minerals |
---|---|
Product Type | Rock and mineral test kit |
Includes | Six specimens, streak plate, plastic dropper bottle, magnet and instructions |
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
S90620
![]() |
Neo/Sci Corporation
203931 |
1 |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Standard tests analyze geological specimens for color, luster, streak, hardness, cleavage and fracture, specific gravity, magnetic attraction and reaction to an acid.
- Perform a variety of tests on the six specimens to identify them as rocks or minerals
- Simple observations and experiments will further classify the rocks as igneous, sedimentary and metamorphic
- Hone observation and analysis skills
Specifications
Used to conduct a series of tests to distinguish between rocks and minerals | |
Rock and mineral test kit | |
Six specimens, streak plate, plastic dropper bottle, magnet and instructions |