Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK1 Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 750, Clone: 261, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242917AF750

Catalog No. NP242917A70


Add to cart

Description

Description

ROCK1 Monoclonal antibody specifically detects ROCK1 in Human,Mouse,Rat samples. It is validated for Western Blot.
Specifications

Specifications

ROCK1
261
Western Blot
EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
ROCK1
Recombinant protein encompassing a sequence within the N-terminus region of human ROCK1. The exact sequence is proprietary.
0.1 mL
Primary
6093
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit