Learn More
ROCK1 Mouse anti-Human, Mouse, Rat, Canine, Primate, Rabbit, Alexa Fluor™ 532, Clone: 154C1465, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10056596AF532
Catalog No. N10696A32
Add to cart
Description
Description
ROCK1 Monoclonal antibody specifically detects ROCK1 in Human,Mouse,Rat,Canine,Primate,Rabbit samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
ROCK1
154C1465
Western Blot, Immunocytochemistry/Immunofluorescence
EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1
Mouse
IgG
Store at 4°C in the dark.
Monoclonal
This mAb recognizes the cleaved C-terminus of ROCK-1. It does not recognize the full-length protein. Since ROCK-1 is cleaved by caspase-3 during apoptosis, this may be used for detection of apoptosis.
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
ROCK1
This antibody was generated by immunizing mice with a synthetic peptide sequence corresponding to amino acids 1114-1126 GNLPESRIEGWLS of human ROCK-1. This sequence is 100% conserved between human, chimpanzee, mouse, rat, dog, and rabbit. This immunogen peptide sequence is adjacent to the caspase-3 cleavage site of ROCK-1.
0.1 mL
Primary
6093
Human, Mouse, Rat, Canine, Primate, Rabbit