Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ROCK1
Applications Immunohistochemistry, Immunoprecipitation, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092625
View Documents
Novus Biologicals
NBP303713100UL
100 μg Each for $320.71
Add to cart
 
NB092626
View Documents
Novus Biologicals
NBP30371320UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ROCK1 Polyclonal antibody specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown
Specifications

Specifications

ROCK1
Unconjugated
Rabbit
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway
PBS with 50% glycerol, pH7.3.
6093
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunoprecipitation, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1
Recombinant fusion protein containing a sequence corresponding to amino acids 400-660 of human ROCK1 (NP_005397.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit