Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROCK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA594955
Description
Synthetic peptide sequence: 35-51aa, RDPRSPINVESLLDGLN. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
The protein encoded by this gene is a serine/threonine kinase that regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. This protein, which is an isozyme of ROCK1 is a target for the small GTPase Rho.Specifications
ROCK2 | |
Polyclonal | |
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl, 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
O75116, P70336, Q62868 | |
ROCK2 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human ROCK2, identical to the related rat and mouse sequences. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ROCK2 | |
Rho-associated protein kinase 2; 2.7.11.1; Rho kinase 2; Rho-associated, coiled-coil-containing protein kinase 2; Rho-associated, coiled-coil-containing protein kinase II; ROCK-II; p164 ROCK-2; ROCK2; KIAA0619 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19878, 25537, 9475 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title