Learn More
Sterlitech CorporationSupplier Diversity Partner Rocker 801, Oil Free Vacuum Pump, AC110V, 60Hz (167801-11) DFS Item
Manufacturer:  Sterlitech CorporationSupplier Diversity Partner 1240163
Horse Power: 1/8, Flow Rate (lpm): 23Vacuum Pump Applications:* Filtration for Microbiology Test* Air Sampling* Vacuum Pad Hold-Down* Suspended Solid Test* Vacuum Filtration
Catalog No. 50-195-2295