Learn More
American Educational Products Rocks and Minerals of U.S. Reference
Excellent reference collections and a valuable aid in identifying and comparing common and rare rocks and minerals
Manufacturer: American Educational Products 2100
Includes: Rocks and minerals study guide
Description
100-Piece Set
- 64 mineral specimens following Dana's System of Mineralogy
- 36 igneous, sedimentary, and metamorphic rocks
- Each approximately 5/8 in.
Order Info
Also available as 24-piece set.
Specifications
Rocks and minerals study guide | |
100 Specimens | |
5/8in. |