Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Mobile Cart/Stationary Table Accessory, Rod Sockets
SCI_ED

Fits tightly into the top of mobile carts and stationary tables and desks

Manufacturer:  Diversified Spaces™ 206505

Catalog No. S94783A


Add to cart

Description

Description

  • Solid aluminum
  • Rust and acid resistant
  • Receive tapered end of 3/4 in. uprights

Order Info

FASTRAK item: ships within 5 days

Specifications

Specifications

1.5 in.
Mobile and stationary tables and desk
2.25 in.
1 lb.
0.45 kg
Rod Socket
3.8 cm
For mobile demonstration units
5.7 cm
1.5 in.
38.1 mm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit