Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROD1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311006100UL
Description
ROD1 Polyclonal specifically detects ROD1 in Mouse samples. It is validated for Western Blot.Specifications
ROD1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
DKFZp781I1117, fission yeast differentiation regulator, PTBP3, regulator of differentiation (in S. pombe) 1, regulator of differentiation 1, Rod1, ROD1 regulator of differentiation 1 (S. pombe) | |
The immunogen is a synthetic peptide directed towards the middle region of mouse ROD1 (NP_659153.2). Peptide sequence QRLYGKVLRATLSKHQAVQLPREGQEDQGLTKDFSNSPLHRFKKPGSKNF | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
9991 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction