Learn More
Selleck Chemical LLCSupplier Diversity Partner Roflumilast 10mg 162401-32-3 APTA 2217, B9302-107, BY 217, BYK 20869  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S213110MG
Roflumilast (APTA 2217, B9302-107, BY 217, BYK 20869) is a selective inhibitor of PDE4 with IC50 of 0.2-4.3 nM in a cell-free assay.
Catalog No. 50-664-8