Learn More
Mopec Roller Autopsy Cart with Fluid Collection System and Chassis Only
Gliding on corrosion proof nylon rollers, the body tray easily rolls onto the cart frame. Mopec Roller Autopsy Cart with Fluid Collection System and Chassis Only is designed with a built in slope and can be used in conjunction with any Mopec wall mounted sinks.
Manufacturer: Mopec HA000
Description
- All welded stainless steel frame, 16 gauge
- Conveyor roller transfer style top
- Optional 23 inch or 27 inch tray with drain hole and plug
- Four heavy duty 8 inch casters with 3 total lock and 1 directional lock
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
Stainless Steel | |
78 in. | |
27 in. | |
88.9cm |
Autopsy Tables and Carts | |
35 in. | |
198.12cm | |
68.58cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.