Learn More
Rolling Collision Balls Products for Science Education
Track and collision balls demonstrate actions and reactions.
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
S40973
View Documents
Sempco Inc
BH0004
N/A Balls Collision Glass Set 6/pk This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Demonstrate the principle that for every action there is an equal and opposite reaction
  • Include a pair of plated steel tracks 31.9 in. (81cm) long and seven steel collision balls 1 in. (2.5cm) in diameter
  • An optional set of six glass collision balls is also available (does not include track)