Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROPN1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548294

Catalog No. PA548294

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and monkey based on sequence homology.

This gene encodes a member of the ropporin family. The encoded protein is present in sperm and interacts with A-kinase anchoring protein, AKAP3, through the amphipathic helix region of AKAP3. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Oct 2014]
Specifications

Specifications

ROPN1L
Polyclonal
PBS with 0.09% sodium azide
Q96C74, Q9EQ00
Ropporin-1-like protein, ROPN1-like protein, AKAP-associated sperm protein, ROPN1L, ASP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
252967, 83853
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ROPN1L
Liquid
ROPN1L
KLH conjugated synthetic peptide between 32-60 amino acids from the N-terminal region of human ROPN1L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit