Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR gamma/RORC/NR1F3 Mouse anti-Human, Clone: RORC/2942, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30756510UG

 View more versions of this product

Catalog No. NB122603


Add to cart

Description

Description

ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human samples. It is validated for ELISA, Protein Array.
Specifications

Specifications

ROR gamma/RORC/NR1F3
RORC/2942
Unconjugated
10 mM PBS with 0.05% BSA
MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR
Recombinant full-length human ROR gamma/RORC/NR1F3 protein
Protein A or G purified
RUO
Primary
6097
ELISA, Peptide Array
0.2 mg/ml
ELISA, Protein Array
Purified
Mouse
IgG2a Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit