Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR gamma/RORC/NR1F3 Mouse anti-Human, Clone: RORC/2942, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ROR gamma/RORC/NR1F3
Clone RORC/2942
Concentration 0.2 mg/ml
Dilution ELISA, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122604
View Documents
Novus Biologicals
NBP30756520UG
20 μg
Each for $138.37
Add to cart
 
NB122603
View Documents
Novus Biologicals
NBP30756510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ROR gamma/RORC/NR1F3 Monoclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

ROR gamma/RORC/NR1F3
0.2 mg/ml
ELISA, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
6097
IgG2a Kappa
Primary
Store at 4°C.
RORC/2942
ELISA, Protein Array
Unconjugated
Mouse
Human
MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR
Recombinant full-length human ROR gamma/RORC/NR1F3 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit