Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR gamma/RORC/NR1F3 Antibody (4G419), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224449AF532
Description
ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
ROR gamma/RORC/NR1F3 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
RORC | |
A portion of amino acids 1-50 of human ROR gamma was used as the immunogen. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Western Blot, Flow Cytometry | |
4G419 | |
Western Blot, Flow Cytometry | |
MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR | |
Mouse | |
Affinity Purified | |
GPCR | |
6097 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction