Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROR gamma/RORC/NR1F3 Mouse anti-Human, Mouse, Rat, Janelia Fluor 646, Clone: 4G419, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP224449JF646
Description
ROR gamma/RORC/NR1F3 Monoclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
ROR gamma/RORC/NR1F3 | |
4G419 | |
Western Blot, Flow Cytometry | |
Affinity Purified, Conjugated | |
RORC | |
A portion of amino acids 1-50 of human ROR gamma was used as the immunogen. | |
Immunogen affinity purified | |
GPCR | |
Primary | |
6097.0 |
Flow Cytometry, Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title