Learn More
ROR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206291AP
Catalog No. 50-173-2999
Add to cart
Description
Description
ROR1 is a receptor protein tyrosine kinase whose cellular role has not been determined. It is a type I membrane protein and belongs to the ROR subfamily of cell surface receptors. Studies of a similar protein in mouse suggest that this protein may interact with another receptor protein tyrosine kinase and may be involved in skeletal and cardiac development.
Specifications
Specifications
ROR1
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q01973, Q9Z139
NTRKR1, ROR1
Rabbit
IgG
150 μL
-20°C
Primary
26563, 362550, 4919
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ROR1
Liquid
Ror1
ROR1 Fusion Protein Ag14586
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat