Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR2 Mouse anti-Human, Clone: ROR2/1912, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ROR2
Clone ROR2/1912
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122398
View Documents
Novus Biologicals
NBP30746220UG
20 μg
Each for $138.37
Add to cart
 
NB122397
View Documents
Novus Biologicals
NBP30746210UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

ROR2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2
Human cultured keratinocytes
Monoclonal
Protein A or G purified
ROR2/1912
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Protein Kinase, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
4920
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit