Learn More
RORB Mouse anti-Human, Clone: OTI4B12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806859
Catalog No. 50-166-8013
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.
Specifications
Specifications
RORB
Monoclonal
Unconjugated
RORB
Lyophilized
RORB
Human recombit protein fragment corresponding to amino acids 1-260 of human RORB produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI4B12
PBS with 8% trehalose and no preservative; pH 7.3
Q92753
NR1F2, ROR-BETA, RZR-BETA, RZRB, bA133M9.1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6096