Learn More
Selleck Chemical LLCSupplier Diversity Partner Rosiglitazone 50mg 122320-73-4 BRL 49653  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S255650MG
Rosiglitazone (BRL 49653) is a potent antihyperglycemic agent and a potent thiazolidinedione insulin sensitizer with IC50 of 12, 4 and 9 nM for rat, 3T3-L1 and human adipocytes, respectively. Rosiglitazone is a pure ligand of PPAR-gamma, and has no PPAR-alpha-binding action. Rosiglitazone modulates TRP channels and induces autophagy. Rosiglitazone prevents ferroptosis.
Catalog No. 50-671-1