Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rosmarinic acid, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
20283-92-5
C18H16O8
360.318
DOUMFZQKYFQNTF-WUTVXBCWSA-N
rosmarinic acid,rosemary acid,r-rosmarinic acid,unii-mqe6xg29yi,rosmarinate,mqe6xg29yi,labiatenic acid,r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid,3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester,2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid
5281792
CHEBI:50371
(2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O
This item is not returnable. View return policy
20283-92-5
C18H16O8
360.318
DOUMFZQKYFQNTF-WUTVXBCWSA-N
rosmarinic acid,rosemary acid,r-rosmarinic acid,unii-mqe6xg29yi,rosmarinate,mqe6xg29yi,labiatenic acid,r-o-3,4-dihydroxycinnamoyl-3-3,4-dihydroxyphenyl lactic acid,3,4-dihydroxycinnamic acid r-1-carboxy-2-3,4-dihydroxyphenyl ethyl ester,2r-3-3,4-dihydroxyphenyl-2-2e-3-3,4-dihydroxyphenyl prop-2-enoyloxy propanoic acid
5281792
CHEBI:50371
(2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
C1=CC(=C(C=C1CC(C(=O)O)OC(=O)C=CC2=CC(=C(C=C2)O)O)O)O

Anti-inflammatory, cytostatic and antiviral; GPR35 agonist

Anti-inflammatory, cytostatic and antiviral. Also displays agonist activity at GPR35.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 360.32
Percent Purity >99%
Chemical Name or Material Rosmarinic acid

Recommended Storage : Store at Room Temperature