Learn More
Selleck Chemical LLCSupplier Diversity Partner Rosuvastatin DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S507210MG
Rosuvastatin is an inhibitor of HMG-CoA reductase, an enzyme that catalyzes the rate-limiting step in cholesterol biosynthesis, with Ki value (inhibition constant) of approximately 0.1 nM.
Catalog No. 50-194-8245